Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018957.3 | NP_061830.3 | 14 | 1354 | ||
GenBank mRNA | AK124370.1 | - | 15 | 2181 | ||
AK126556.1 | - | 13 | 2049 | |||
AK301163.1 | - | 14 | 1336 | |||
AL157480.1 | - | 3 | 178 | |||
AY364241.1 | - | 11 | 1054 | |||
BC008282.1 | - | 12 | 1147 | 3639782 | ||
CR456576.1 | - | 14 | 1273 |
Gene Symbol: LOC101927051 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109952.1 | 4 | 634 | |||
GenBank mRNA | AK097791.1 | - | 5 | 634 |