Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256732.1 | NP_001243661.1 | 16 | 1240 | ||
NM_001256733.1 | NP_001243662.1 | 15 | 1156 | |||
NM_001256734.1 | NP_001243663.1 | 15 | 1150 | |||
NM_001256735.1 | NP_001243664.1 | 15 | 1126 | |||
NM_012446.3 | NP_036578.2 | 16 | 1216 | |||
XM_017009304.1 | XP_016864793.1 | 16 | 1058 | |||
XM_017009305.1 | XP_016864794.1 | 17 | 1147 | |||
XM_017009307.1 | XP_016864796.1 | 15 | 944 | |||
XM_017009308.1 | XP_016864797.1 | 17 | 1319 | |||
XM_017009309.1 | XP_016864798.1 | 16 | 1383 | |||
XM_017009310.1 | XP_016864799.1 | 16 | 1531 | |||
GenBank mRNA | AF077048.1 | - | 16 | 1080 | ||
AF161465.1 | - | 16 | 1046 | |||
AK074537.1 | - | 16 | 1011 | |||
AK293544.1 | - | 15 | 951 | |||
AK297494.1 | - | 16 | 924 | |||
AK299858.1 | - | 15 | 994 | |||
AK312337.1 | - | 16 | 1060 | |||
AL080076.1 | - | 16 | 1064 | |||
BC017020.1 | - | 16 | 1064 | 3913337 | ||
BX397789.2 | - | 2 | 679 | |||
CR936787.1 | - | 15 | 971 | |||
DA202987.1 | - | 8 | 406 |