Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012134.2 | NP_036266.2 | 2, 3 | 2025 | ||
GenBank mRNA | AK126474.1 | - | 2, 3 | 1564 | ||
AK295824.1 | - | 2, 3 | 1931 | |||
AK304700.1 | - | 2, 3 | 1642 | |||
AK304852.1 | - | 3, 4 | 1791 | |||
BC001755.1 | - | 3, 4 | 997 | |||
BC080187.1 | - | 2, 3 | 1974 | 6177902 | ||
X54162.1 | - | 2, 3 | 1905 |