Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AFF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014423.3 | NP_055238.1 | 3 | 1077 | ||
XM_005271963.4 | XP_005272020.1 | 4 | 1141 | |||
XM_006714587.3 | XP_006714650.1 | 3 | 1077 | |||
GenBank mRNA | AB209547.1 | - | 3 | 1033 | ||
AF197927.1 | - | 3 | 1006 | |||
AF213987.2 | - | 3 | 734 | |||
AK024831.1 | - | 1 | 535 | |||
AK055638.1 | - | 3 | 1025 | |||
AK307407.1 | - | 3 | 1077 | |||
BC025700.1 | - | 3 | 929 | |||
BC063007.1 | - | 3 | 928 | |||
BC100287.1 | - | 3 | 1029 | |||
BC137226.1 | - | 3 | 975 | 9021223 | ||
BC144538.1 | - | 3 | 975 |