Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPNE7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014427.4 | NP_055242.1 | 11 | 1240 | ||
NM_153636.2 | NP_705900.1 | 9 | 1015 | |||
XM_011523000.1 | XP_011521302.1 | 9 | 1010 | |||
XM_011523001.2 | XP_011521303.1 | 9 | 775 | |||
XM_017023138.1 | XP_016878627.1 | 11 | 1204 | |||
XM_017023139.1 | XP_016878628.1 | 11 | 1215 | |||
XM_017023140.1 | XP_016878629.1 | 9 | 990 | |||
XM_017023141.1 | XP_016878630.1 | 8 | 2097 | |||
GenBank mRNA | AJ133798.1 | - | 11 | 1239 | ||
BC035334.1 | - | 9 | 953 | 5191900 | ||
BC064577.1 | - | 9 | 992 | 5727324 |