Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199835.1 | NP_001186764.1 | 5 | 558 | ||
NM_001199837.1 | NP_001186766.1 | 3 | 363 | |||
NM_001199838.1 | NP_001186767.1 | 2 | 262 | |||
NM_001318198.1 | NP_001305127.1 | 6 | 632 | |||
NM_001318199.1 | NP_001305128.1 | 5 | 433 | |||
NM_013322.2 | NP_037454.2 | 5 | 511 | |||
XM_006715712.2 | XP_006715775.1 | 5 | 711 | |||
XM_017012085.1 | XP_016867574.1 | 7 | 726 | |||
XM_017012086.1 | XP_016867575.1 | 7 | 1165 | |||
XM_017012087.1 | XP_016867576.1 | 6 | 655 | |||
XM_017012088.1 | XP_016867577.1 | 6 | 602 | |||
GenBank mRNA | AF121860.1 | - | 5 | 424 | ||
AK296142.1 | - | 6 | 462 | |||
AK307905.1 | - | 3 | 312 | |||
AK309162.1 | - | 2 | 262 | |||
AK312850.1 | - | 5 | 433 | |||
AK316088.1 | - | 6 | 608 | |||
BC031050.1 | - | 5 | 405 | 5278015 | ||
BC034992.1 | - | 5 | 484 | 4827599 | ||
BJ995940.1 | - | 3 | 363 | |||
DA377368.1 | - | 5 | 433 |