Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STOML2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287032.1 | NP_001273961.1 | 6 | 515 | ||
NM_001287033.1 | NP_001273962.1 | 5 | 488 | |||
NM_013442.2 | NP_038470.1 | 6 | 626 | |||
GenBank mRNA | AF161458.1 | - | 6 | 500 | ||
AF190167.1 | - | 6 | 588 | |||
AF282596.1 | - | 6 | 559 | |||
AK027405.1 | - | 6 | 559 | |||
AK130546.1 | - | 3 | 1406 | |||
AK223102.1 | - | 6 | 575 | |||
AK225114.1 | - | 6 | 559 | |||
BC002442.2 | - | 6 | 562 | 3346384 | ||
BC003025.1 | - | 6 | 547 | 2821269 | ||
BC010152.2 | - | 6 | 559 | 3534656 | ||
BC014990.1 | - | 6 | 557 | 4907745 | ||
BQ935988.1 | - | 6 | 503 | |||
BU528995.1 | - | 5 | 385 | |||
BX400542.2 | - | 6 | 513 |