Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAS2R13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023920.2 | NP_076409.1 | 1 | 443 | ||
GenBank mRNA | BC069376.1 | - | 1 | 179 | 7262240 | |
BC095518.1 | - | 1 | 443 | 30706594 | ||
BC115402.1 | - | 1 | 172 |