Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DERL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304777.1 | NP_001291706.1 | 6 | 644 | ||
NM_001304779.1 | NP_001291708.1 | 3 | 283 | |||
NM_016041.4 | NP_057125.2 | 6 | 647 | |||
NR_130905.1 | 5 | 759 | ||||
NR_130906.1 | 5 | 553 | ||||
GenBank mRNA | AF132289.1 | - | 6 | 595 | ||
AF151859.1 | - | 6 | 585 | |||
AF208065.1 | - | 6 | 583 | |||
AF242523.1 | - | 6 | 583 | |||
AK024574.1 | - | 2 | 265 | |||
BC010890.1 | - | 6 | 595 | 4280273 | ||
BU199049.1 | - | 3 | 215 | |||
CN362881.1 | - | 6 | 565 | |||
CN362882.1 | - | 5 | 471 | |||
CR457202.1 | - | 6 | 578 |