Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SSUH2 SIRNA Assays ›
Gene Symbol:
SSUH2
Gene Name:
ssu-2 homolog (C. elegans)
Gene Aliases:
C3orf32, SSU-2, fls485
Chromosome Location:
Chr.3: 8619400 - 8745035 on Build GRCh38
UniGene ID:
Hs.736935
Species:
Human
siRNA ID
HSS121573
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SSUH2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256748.1
NP_001243677.1
12
1089
NM_001256749.1
NP_001243678.1
12
1110
NM_015931.2
NP_057015.1
12
1256
NR_046358.1
13
1337
XM_011533774.1
XP_011532076.1
20
2805
XM_017006510.1
XP_016861999.1
13
5570
XM_017006511.1
XP_016862000.1
19
2493
XM_017006512.1
XP_016862001.1
19
2662
XM_017006513.1
XP_016862002.1
18
2498
XM_017006514.1
XP_016862003.1
18
2404
XM_017006515.1
XP_016862004.1
20
3559
XM_017006516.1
XP_016862005.1
15
1920
XM_017006517.1
XP_016862006.1
15
1873
XM_017006518.1
XP_016862007.1
20
2784
XM_017006519.1
XP_016862008.1
13
5515
XM_017006520.1
XP_016862009.1
13
1866
XM_017006521.1
XP_016862010.1
16
2024
XM_017006522.1
XP_016862011.1
16
1915
XM_017006523.1
XP_016862012.1
14
1618
XM_017006524.1
XP_016862013.1
15
1856
XM_017006525.1
XP_016862014.1
18
2425
XM_017006526.1
XP_016862015.1
13
1504
XM_017006527.1
XP_016862016.1
19
2547
XM_017006528.1
XP_016862017.1
13
1543
XM_017006529.1
XP_016862018.1
13
1573
XM_017006530.1
XP_016862019.1
15
2006
XM_017006531.1
XP_016862020.1
14
1928
XM_017006533.1
XP_016862022.1
19
2580
XM_017006534.1
XP_016862023.1
19
2526
XM_017006538.1
XP_016862027.1
17
2123
XM_017006539.1
XP_016862028.1
12
1235
XM_017006540.1
XP_016862029.1
13
1414
XM_017006541.1
XP_016862030.1
10
1056
XM_017006542.1
XP_016862031.1
10
1034
GenBank mRNA
AB024705.1
-
12
1229
AK057972.1
-
13
1333
AK093025.1
-
1
1798
AK093066.1
-
18
2286
AK122938.1
-
17
2854
AK225925.1
-
13
1333
AK300175.1
-
12
1089
BC052614.1
-
12
1093
6300057
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-f6n66:80/10.239.161.116:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0