Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTFDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001283035.1 | NP_001269964.1 | 3 | 306 | ||
NM_001283036.1 | NP_001269965.1 | 2 | 216 | |||
NM_001283037.1 | NP_001269966.1 | 2 | 216 | |||
NM_016407.4 | NP_057491.2 | 2 | 216 | |||
XM_017027872.1 | XP_016883361.1 | 5 | 409 | |||
GenBank mRNA | AF117231.1 | - | 2 | 164 | ||
AF161513.1 | - | 2 | 173 | |||
AF161518.1 | - | 2 | 142 | |||
AF212244.1 | - | 2 | 153 | |||
AK000481.1 | - | 2 | 146 | |||
AK026118.1 | - | 2 | 144 | |||
AK225135.1 | - | 2 | 134 | |||
AK293662.1 | - | 2 | 144 | |||
AK302029.1 | - | 3 | 249 | |||
AK314563.1 | - | 2 | 171 | |||
BC003359.1 | - | 2 | 143 | 2900332 | ||
BP307634.1 | - | 2 | 210 | |||
BU537939.1 | - | 2 | 126 | |||
CR983692.1 | - | 2 | 127 | |||
DC397043.1 | - | 3 | 306 |