Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SASH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018990.3 | NP_061863.1 | 3 | 422 | ||
XM_006724763.1 | XP_006724826.1 | 3 | 389 | |||
GenBank mRNA | AK296665.1 | - | 3 | 324 | ||
AK314869.1 | - | 3 | 389 | |||
AL049683.1 | - | 3 | 347 | |||
BC051881.1 | - | 3 | 354 | 6011968 | ||
CR975883.1 | - | 3 | 422 |