Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other UCKL1 SIRNA Assays ›
Gene Symbol:
UCKL1
Gene Name:
uridine-cytidine kinase 1 like 1
Gene Aliases:
UCK1L, URKL1
Chromosome Location:
Chr.20: 63939829 - 63959435 on Build GRCh38
UniGene ID:
Hs.504998
Species:
Human
siRNA ID
HSS123624
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: UCKL1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001193379.1
NP_001180308.1
6
1055
NM_017859.3
NP_060329.2
6
827
NR_126526.1
6
827
XM_005260216.2
XP_005260273.1
6
795
XM_006723806.2
XP_006723869.1
6
795
XM_006723807.2
XP_006723870.1
6
795
XM_006723808.1
XP_006723871.1
6
847
XM_006723809.1
XP_006723872.1
6
827
XM_006723810.1
XP_006723873.1
6
824
XM_006723811.3
XP_006723874.1
6
2816
XM_011528868.2
XP_011527170.1
6
2819
XM_011528869.1
XP_011527171.1
7
4090
XM_011528870.1
XP_011527172.1
6
809
XM_011528871.1
XP_011527173.1
6
836
XM_011528872.1
XP_011527174.1
7
4079
XM_011528873.2
XP_011527175.1
6
795
XM_017027894.1
XP_016883383.1
6
795
XM_017027895.1
XP_016883384.1
6
795
XM_017027896.1
XP_016883385.1
6
827
XM_017027897.1
XP_016883386.1
6
827
XM_017027898.1
XP_016883387.1
6
824
XM_017027899.1
XP_016883388.1
6
824
XM_017027900.1
XP_016883389.1
6
1673
XM_017027901.1
XP_016883390.1
6
1671
XM_017027902.1
XP_016883391.1
7
1027
XM_017027903.1
XP_016883392.1
6
836
XM_017027904.1
XP_016883393.1
6
3751
XM_017027905.1
XP_016883394.1
6
3754
XM_017027906.1
XP_016883395.1
6
3751
XM_017027907.1
XP_016883396.1
2
547
XM_017027908.1
XP_016883397.1
3
450
GenBank mRNA
AJ605558.1
-
6
796
AK000524.1
-
6
796
AK128082.1
-
1
2141
AK222559.1
-
6
798
AK223220.1
-
6
796
AK301602.1
-
6
735
AK303688.1
-
6
1007
BC033078.1
-
6
765
4810505
Back To Top
More Information
Panther Classification:
Molecular Function -
nucleotide kinase
Gene Ontology Categories:
Function(s)
Process(es)
pyrimidine nucleobase metabolic process
viral process
phosphorylation
pyrimidine nucleoside salvage
UMP salvage
CTP salvage
uridine kinase activity
protein binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-9fjjm:80/10.239.165.64:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0