Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318533.1 | NP_001305462.1 | 21 | 2040 | ||
NM_017868.3 | NP_060338.3 | 21 | 2022 | |||
XM_005271604.3 | XP_005271661.1 | 21 | 2238 | |||
XM_011542890.2 | XP_011541192.1 | 22 | 2236 | |||
XM_017017943.1 | XP_016873432.1 | 21 | 2022 | |||
XM_017017944.1 | XP_016873433.1 | 22 | 2592 | |||
XM_017017945.1 | XP_016873434.1 | 22 | 2075 | |||
XM_017017946.1 | XP_016873435.1 | 21 | 2245 | |||
XM_017017947.1 | XP_016873436.1 | 21 | 2022 | |||
XM_017017948.1 | XP_016873437.1 | 21 | 2022 | |||
XM_017017949.1 | XP_016873438.1 | 21 | 2022 | |||
XM_017017950.1 | XP_016873439.1 | 21 | 2022 | |||
GenBank mRNA | AK000542.1 | - | 20 | 1914 | ||
AK023921.1 | - | 21 | 1968 | |||
AK091887.1 | - | 23 | 2634 | |||
AK223117.1 | - | 20 | 1914 | |||
AK292331.1 | - | 21 | 1960 | |||
BC011546.1 | - | 20 | 1964 | |||
BC032355.1 | - | 20 | 1944 | 5219117 |