Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PLEKHB2 SIRNA Assays ›
Gene Symbol:
PLEKHB2
Gene Name:
pleckstrin homology domain containing B2
Gene Aliases:
EVT2
Chromosome Location:
Chr.2: 131104847 - 131149852 on Build GRCh38
UniGene ID:
Hs.469944
Species:
Human
siRNA ID
HSS123790
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PLEKHB2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001100623.1
NP_001094093.1
7
1028
NM_001267062.1
NP_001253991.1
7
1028
NM_001267063.1
NP_001253992.1
7
1025
NM_001267064.1
NP_001253993.1
7
1049
NM_001267065.1
NP_001253994.1
7
1052
NM_001267067.1
NP_001253996.1
5
898
NM_001267068.1
NP_001253997.1
5
895
NM_001309448.1
NP_001296377.1
6
983
NM_001309450.1
NP_001296379.1
5
885
NM_001309451.1
NP_001296380.1
6
980
NM_001309452.1
NP_001296381.1
8
1126
NM_017958.2
NP_060428.2
7
1025
XM_011511385.2
XP_011509687.1
7
591
XM_011511386.2
XP_011509688.1
7
591
XM_017004408.1
XP_016859897.1
6
520
XM_017004409.1
XP_016859898.1
6
496
XM_017004410.1
XP_016859899.1
6
520
XM_017004411.1
XP_016859900.1
8
1656
XM_017004412.1
XP_016859901.1
8
1632
XM_017004413.1
XP_016859902.1
5
416
GenBank mRNA
AK000790.1
-
7
536
AK093730.1
-
7
1263
AK123710.1
-
7
985
AK126289.1
-
7
1052
AK223338.1
-
7
536
AK296008.1
-
6
492
AK302770.1
-
5
440
BC013991.2
-
7
521
3639839
BC050695.1
-
7
523
6162960
BP362174.1
-
6
491
BQ052811.1
-
7
520
BQ649140.1
-
7
558
BU174451.1
-
8
607
CK000843.1
-
5
388
DB294071.1
-
7
538
DC303684.1
-
5
416
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cell differentiation
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-zp8kv:80/10.239.163.191:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0