Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCUN1D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014283.1 | NP_001014305.1 | 3 | 292 | ||
XM_017020643.1 | XP_016876132.1 | 3 | 503 | |||
XM_017020644.1 | XP_016876133.1 | 3 | 433 | |||
XM_017020645.1 | XP_016876134.1 | 3 | 474 | |||
XM_017020646.1 | XP_016876135.1 | 3 | 4215 | |||
XM_017020647.1 | XP_016876136.1 | 3 | 753 | |||
XM_017020648.1 | XP_016876137.1 | 3 | 1516 | |||
XM_017020649.1 | XP_016876138.1 | 2 | 330 | |||
GenBank mRNA | AK001566.1 | - | 3 | 275 | ||
BC056669.1 | - | 3 | 279 | 6494926 | ||
BM836766.1 | - | 3 | 435 |