Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF280C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017666.4 | NP_060136.1 | 15, 16 | 2192 | ||
XM_006724765.3 | XP_006724828.1 | 14, 15 | 1996 | |||
GenBank mRNA | AK290373.1 | - | 15, 16 | 2143 | ||
AL834333.1 | - | 15, 16 | 2123 | |||
BC051728.1 | - | 15, 16 | 2143 | 4828435 | ||
DA441065.1 | - | 2, 3 | 216 |