Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POMGNT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243766.1 | NP_001230695.1 | 20 | 2377 | ||
NM_001290129.1 | NP_001277058.1 | 19 | 1798 | |||
NM_001290130.1 | NP_001277059.1 | 19 | 1764 | |||
NM_017739.3 | NP_060209.3 | 20 | 1890 | |||
XM_005271010.1 | XP_005271067.1 | 20 | 1885 | |||
XM_006710755.1 | XP_006710818.1 | 20 | 1850 | |||
XM_006710756.1 | XP_006710819.1 | 20 | 1885 | |||
XM_011541760.2 | XP_011540062.1 | 19 | 1742 | |||
XM_017001690.1 | XP_016857179.1 | 20 | 1850 | |||
GenBank mRNA | AB057356.1 | - | 20 | 1820 | ||
AF250859.2 | - | 17 | 1479 | |||
AK000284.1 | - | 20 | 1867 | |||
AK022727.1 | - | 15 | 1236 | |||
AK056186.1 | - | 20 | 2377 | |||
AK225033.1 | - | 20 | 1868 | |||
AK301923.1 | - | 19 | 1764 | |||
AK302379.1 | - | 19 | 1774 | |||
AK316399.1 | - | 19 | 1771 | |||
AW818474.1 | - | 2 | 159 | |||
AY358592.1 | - | 20 | 1855 | |||
BC001471.2 | - | 20 | 1884 | 3138971 | ||
CR749700.1 | - | 17 | 2635 |