Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTBD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017797.3 | NP_060267.2 | 7 | 1203 | ||
XM_005259593.1 | XP_005259650.1 | 7 | 968 | |||
XM_011528127.1 | XP_011526429.1 | 5 | 569 | |||
GenBank mRNA | AF355797.1 | - | 7 | 1203 | ||
AK000393.1 | - | 4 | 997 | |||
AK027481.1 | - | 5 | 565 | |||
AK056818.1 | - | 1 | 1338 | |||
AK225831.1 | - | 7 | 1202 | |||
AL360275.1 | - | 4 | 434 | |||
AY007116.1 | - | 7 | 925 | |||
BC000564.2 | - | 4 | 493 | |||
BC008035.2 | - | 4 | 487 | |||
BC053986.1 | - | 3 | 388 | |||
BC069199.1 | - | 5 | 629 | |||
BC094820.1 | - | 7 | 937 | |||
CR457230.1 | - | 3 | 240 |