Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018238.3 | NP_060708.1 | 8 | 651 | ||
XM_005250023.3 | XP_005250080.1 | 8 | 547 | |||
XM_011516397.2 | XP_011514699.1 | 8 | 1117 | |||
GenBank mRNA | AJ278150.1 | - | 7 | 507 | ||
AK001704.1 | - | 8 | 519 | |||
AK222769.1 | - | 8 | 513 | |||
AK299131.1 | - | 8 | 510 | |||
AK304502.1 | - | 9 | 538 | |||
BC022777.1 | - | 7 | 505 | 3913448 |