Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERO1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019891.3 | NP_063944.3 | 15 | 1532 | ||
GenBank mRNA | AF252538.1 | - | 15 | 1449 | ||
AK315351.1 | - | 15 | 1451 | |||
BC032823.2 | - | 15 | 1383 | 4829502 | ||
BC044573.1 | - | 15 | 1478 | 5270105 |