Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020154.2 | NP_064539.1 | 1 | 234 | ||
GenBank mRNA | AF242729.1 | - | 1 | 271 | ||
AJ245874.1 | - | 1 | 256 | |||
AJ250344.1 | - | 1 | 234 | |||
AK314883.1 | - | 1 | 303 | |||
AY358445.1 | - | 1 | 252 | |||
BC012456.1 | - | 1 | 198 | |||
BC104934.1 | - | 1 | 312 | 8143937 | ||
BC104936.1 | - | 1 | 312 | 8143939 |