Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082533.1 | NP_001076002.1 | 8 | 1871 | ||
NM_001082534.1 | NP_001076003.1 | 8 | 1436 | |||
NM_020178.4 | NP_064563.1 | 7 | 1745 | |||
XM_017024878.1 | XP_016880367.1 | 5 | 599 | |||
GenBank mRNA | AB001597.1 | - | 7 | 759 | ||
AB036836.1 | - | 8 | 1661 | |||
AF288385.1 | - | 8 | 1427 | |||
AK294652.1 | - | 6 | 1116 | |||
AK313001.1 | - | 7 | 1321 | |||
AL161812.1 | - | 7 | 761 | |||
AY358509.1 | - | 7 | 1039 | |||
BC020577.1 | - | 8 | 1403 | 4181377 | ||
BC029865.1 | - | 8 | 1405 | |||
BC047456.1 | - | 7 | 1658 | 5276935 | ||
BC068462.1 | - | 7 | 1704 | 5260321 | ||
CR749607.1 | - | 8 | 1207 |