Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XAB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020196.2 | NP_064581.2 | 10 | 1291 | ||
GenBank mRNA | AB026111.1 | - | 10 | 1280 | ||
AB033003.1 | - | 8 | 967 | |||
AF258567.1 | - | 10 | 1286 | |||
AF272147.1 | - | 10 | 1301 | |||
AK025858.1 | - | 5 | 574 | |||
AK074035.1 | - | 6 | 1862 | |||
BC007208.1 | - | 10 | 1273 | 2823235 | ||
BC008778.1 | - | 5 | 606 | |||
CR749864.1 | - | 7 | 820 |