Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278596.1 | NP_001265525.1 | 20 | 2007 | ||
NM_001278597.1 | NP_001265526.1 | 20 | 2025 | |||
XM_006718988.3 | XP_006719051.1 | 18 | 2282 | |||
XM_011520975.2 | XP_011519277.1 | 18 | 4568 | |||
XM_011520978.2 | XP_011519280.1 | 17 | 1880 | |||
XM_017019672.1 | XP_016875161.1 | 18 | 2282 | |||
GenBank mRNA | AJ272204.1 | - | 20 | 2025 | ||
AK024010.1 | - | 3 | 289 | |||
FJ384095.1 | - | 20 | 1981 |