Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LYRM1 SIRNA Assays ›
Gene Symbol:
LYRM1
Gene Name:
LYR motif containing 1
Gene Aliases:
A211C6.1
Chromosome Location:
Chr.16: 20899626 - 20925006 on Build GRCh38
UniGene ID:
Hs.745012
Species:
Human
siRNA ID
HSS126067
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LYRM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001128301.2
NP_001121773.1
4
675
NM_001128302.2
NP_001121774.1
3
443
NM_001302835.1
NP_001289764.1
5
619
NM_001302836.1
NP_001289765.1
5
771
NM_020424.4
NP_065157.1
4
594
NR_126516.1
3
516
NR_126520.1
2
298
NR_126521.1
2
284
NR_126527.1
2
112
NR_126528.1
3
530
XM_005255444.1
XP_005255501.1
4
496
XM_011545900.2
XP_011544202.1
4
258
XM_011545901.2
XP_011544203.1
4
324
XM_011545902.1
XP_011544204.1
5
595
XM_011545908.2
XP_011544210.1
4
326
XM_017023475.1
XP_016878964.1
5
1011
XM_017023477.1
XP_016878966.1
3
846
XM_017023478.1
XP_016878967.1
4
316
XM_017023479.1
XP_016878968.1
4
753
XM_017023480.1
XP_016878969.1
6
839
XM_017023481.1
XP_016878970.1
4
468
XM_017023482.1
XP_016878971.1
3
522
GenBank mRNA
AK308690.1
-
2
363
AK310055.1
-
3
344
AK311321.1
-
4
496
AK311415.1
-
3
518
AK311881.1
-
3
310
AV714321.1
-
3
289
BC017039.1
-
5
619
3849248
BF575136.1
-
3
332
BG541469.1
-
2
147
BI547471.1
-
3
361
BI825882.1
-
5
618
BM546065.1
-
2
248
BX096767.1
-
2
112
CD237780.1
-
3
313
CX163936.1
-
3
320
DA572182.1
-
4
560
DB054440.1
-
4
531
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-c74kb:80/10.239.167.51:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0