Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LYRM1 SIRNA Assays ›
Gene Symbol:
LYRM1
Gene Name:
LYR motif containing 1
Gene Aliases:
A211C6.1
Chromosome Location:
Chr.16: 20899626 - 20925006 on Build GRCh38
UniGene ID:
Hs.745012
Species:
Human
siRNA ID
HSS126069
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LYRM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001128301.2
NP_001121773.1
5
781
NM_001128302.2
NP_001121774.1
4
549
NM_001302835.1
NP_001289764.1
6
725
NM_001302836.1
NP_001289765.1
6
877
NM_020424.4
NP_065157.1
5
700
NR_126516.1
4
622
NR_126520.1
3
404
NR_126521.1
3
390
NR_126527.1
3
218
NR_126528.1
4
636
XM_005255444.1
XP_005255501.1
5
602
XM_011545900.2
XP_011544202.1
5
364
XM_011545901.2
XP_011544203.1
5
430
XM_011545902.1
XP_011544204.1
6
701
XM_011545908.2
XP_011544210.1
5
432
XM_017023475.1
XP_016878964.1
6
1117
XM_017023476.1
XP_016878965.1
4
1015
XM_017023477.1
XP_016878966.1
4
952
XM_017023478.1
XP_016878967.1
5
422
XM_017023479.1
XP_016878968.1
5
859
XM_017023480.1
XP_016878969.1
7
945
XM_017023481.1
XP_016878970.1
5
574
XM_017023482.1
XP_016878971.1
4
628
GenBank mRNA
AK308690.1
-
3
469
AK311415.1
-
4
624
AK311881.1
-
4
416
AV714321.1
-
4
395
BC017039.1
-
6
725
3849248
BG541469.1
-
3
253
BI825882.1
-
6
724
BM546065.1
-
3
354
BX096767.1
-
3
218
CD237780.1
-
4
419
CX163936.1
-
4
426
DA572182.1
-
5
666
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-7dvbj:80/10.239.167.51:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0