Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL22RA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021258.3 | NP_067081.2 | 7 | 1484 | ||
XM_011541882.1 | XP_011540184.1 | 8 | 1433 | |||
GenBank mRNA | AF286095.1 | - | 7 | 1304 | ||
AK226001.1 | - | 8 | 1245 | |||
AK292204.1 | - | 7 | 1379 | |||
AK304454.1 | - | 5 | 1229 | |||
AK313971.1 | - | 7 | 1484 | |||
BC029273.1 | - | 7 | 1320 | 4347189 |