Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EVA1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286556.1 | NP_001273485.1 | 7 | 1306 | ||
NM_001320744.1 | NP_001307673.1 | 5 | 1050 | |||
NM_001320745.1 | NP_001307674.1 | 7 | 1240 | |||
NM_058187.4 | NP_478067.2 | 7 | 1315 | |||
NR_104472.1 | 6 | 1109 | ||||
NR_135467.1 | 7 | 1414 | ||||
XM_006724038.3 | XP_006724101.1 | 7 | 1433 | |||
XM_011529669.2 | XP_011527971.1 | 7 | 1567 | |||
XM_017028418.1 | XP_016883907.1 | 7 | 1433 | |||
XM_017028419.1 | XP_016883908.1 | 6 | 1355 | |||
XM_017028420.1 | XP_016883909.1 | 8 | 1356 | |||
XM_017028421.1 | XP_016883910.1 | 7 | 1231 | |||
XM_017028422.1 | XP_016883911.1 | 5 | 1106 | |||
XM_017028423.1 | XP_016883912.1 | 4 | 1034 | |||
GenBank mRNA | AF358258.1 | - | 7 | 898 | ||
AK125013.1 | - | 6 | 1865 | |||
AW957938.1 | - | 2 | 127 | |||
AY040087.1 | - | 6 | 701 | |||
AY358787.1 | - | 7 | 1308 | |||
BC038710.1 | - | 7 | 1041 | 4813873 | ||
BX342316.2 | - | 2 | 586 | |||
BX373211.1 | - | 2 | 612 | |||
CA412371.1 | - | 2 | 646 |