Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HRH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021624.3 | NP_067637.2 | 2 | 330 | ||
XM_011526133.1 | XP_011524435.1 | 2 | 391 | |||
GenBank mRNA | AB044934.1 | - | 2 | 284 | ||
AB045370.1 | - | 2 | 293 | |||
AF307973.1 | - | 2 | 230 | |||
AF312230.1 | - | 2 | 330 | |||
AF325356.1 | - | 2 | 230 | |||
AF329449.1 | - | 2 | 230 | |||
AJ298292.1 | - | 2 | 230 | |||
AY008280.1 | - | 2 | 230 | |||
AY136745.1 | - | 2 | 230 | |||
BC069136.1 | - | 2 | 330 | 7216992 | ||
BC112348.1 | - | 2 | 290 | 40006540 |
Set Membership: |
G-protein coupled receptor |