Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALOXE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165960.1 | NP_001159432.1 | 15 | 2460 | ||
NM_021628.2 | NP_067641.2 | 15 | 2296 | |||
GenBank mRNA | AF182218.1 | - | 14 | 2127 | ||
AJ269499.1 | - | 14 | 2425 | |||
AK292732.1 | - | 14 | 2199 | |||
AK296416.1 | - | 15 | 2361 | |||
AK313677.1 | - | 15 | 2260 | |||
BC101938.1 | - | 14 | 2148 | 40011855 | ||
BC101939.1 | - | 14 | 2148 | 40011856 | ||
BC103508.1 | - | 14 | 2148 | 40011857 | ||
BC104724.1 | - | 14 | 2148 | 40011849 |