Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRAGC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271851.1 | NP_001258780.1 | 7 | 1251 | ||
NM_022157.3 | NP_071440.1 | 7 | 1353 | |||
GenBank mRNA | AF272035.1 | - | 7 | 1181 | ||
AF323609.1 | - | 7 | 1176 | |||
AK023373.1 | - | 3 | 381 | |||
AK092808.1 | - | 2 | 2264 | |||
AK298573.1 | - | 6 | 876 | |||
AK298788.1 | - | 7 | 1147 | |||
AK315791.1 | - | 7 | 1277 | |||
BC016668.1 | - | 7 | 1198 | 4342448 | ||
CA449149.1 | - | 1 | 243 | |||
CX761032.1 | - | 2 | 263 |