Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011543544.1 | XP_011541846.1 | 12 | 2043 | ||
GenBank mRNA | AB109031.1 | - | 13 | 2079 | ||
AB163917.1 | - | 13 | 2135 | |||
AF191545.1 | - | 12 | 2051 | |||
AK074281.1 | - | 13 | 2156 | |||
AK074387.1 | - | 6 | 660 | |||
AK312864.1 | - | 13 | 2092 | |||
AY028805.1 | - | 12 | 2065 | |||
BC065240.1 | - | 12 | 1938 | 6166289 |