Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127235.2 | NP_001120707.2 | 2 | 1033 | ||
NM_001127236.2 | NP_001120708.1 | 1 | 80 | |||
NM_022913.3 | NP_075064.1 | 3 | 1297 | |||
XM_006714673.3 | XP_006714736.1 | 3 | 1352 | |||
XM_006714674.3 | XP_006714737.1 | 3 | 1352 | |||
XM_017009754.1 | XP_016865243.1 | 2 | 705 | |||
XM_017009755.1 | XP_016865244.1 | 2 | 705 | |||
XM_017009756.1 | XP_016865245.1 | 2 | 705 | |||
XM_017009757.1 | XP_016865246.1 | 3 | 1352 | |||
XM_017009758.1 | XP_016865247.1 | 3 | 1352 | |||
XM_017009759.1 | XP_016865248.1 | 3 | 1149 | |||
GenBank mRNA | AL136844.1 | - | 1 | 33 | ||
AY226828.1 | - | 3 | 1128 | |||
BC000267.1 | - | 2 | 535 | |||
BC113004.2 | - | 2 | 1046 | 40034860 |