Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127235.2 | NP_001120707.2 | 7 | 1544 | ||
NM_001127236.2 | NP_001120708.1 | 6 | 591 | |||
NM_001203246.1 | NP_001190175.1 | 6 | 1667 | |||
NM_022913.3 | NP_075064.1 | 7 | 1787 | |||
XM_006714673.3 | XP_006714736.1 | 9 | 1923 | |||
XM_006714674.3 | XP_006714737.1 | 8 | 1902 | |||
XM_017009754.1 | XP_016865243.1 | 8 | 1276 | |||
XM_017009755.1 | XP_016865244.1 | 7 | 1255 | |||
XM_017009756.1 | XP_016865245.1 | 7 | 1216 | |||
XM_017009757.1 | XP_016865246.1 | 8 | 1863 | |||
XM_017009758.1 | XP_016865247.1 | 7 | 1842 | |||
XM_017009759.1 | XP_016865248.1 | 7 | 1639 | |||
GenBank mRNA | AK024807.1 | - | 1 | 26 | ||
AK307505.1 | - | 5 | 440 | |||
AL136844.1 | - | 6 | 544 | |||
AL161991.1 | - | 6 | 1566 | |||
AY226828.1 | - | 7 | 1618 | |||
BC000267.1 | - | 7 | 1046 | |||
BC070132.1 | - | 3 | 270 | |||
BC113004.2 | - | 6 | 1536 | 40034860 | ||
CD657101.1 | - | 5 | 436 |