Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CAPRIN2 SIRNA Assays ›
Gene Symbol:
CAPRIN2
Gene Name:
caprin family member 2
Gene Aliases:
C1QDC1, EEG-1, EEG1, RNG140
Chromosome Location:
Chr.12: 30709552 - 30754994 on Build GRCh38
UniGene ID:
Hs.234355
Species:
Human
siRNA ID
HSS128055
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CAPRIN2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001002259.2
NP_001002259.1
18
3936
NM_001206856.2
NP_001193785.1
17
3812
NM_001319842.1
NP_001306771.1
17
3850
NM_001319843.1
NP_001306772.1
18
3933
NM_001319844.1
NP_001306773.1
18
3086
NM_001319845.1
NP_001306774.1
19
3191
NM_001319846.1
NP_001306775.1
19
3181
NM_023925.4
NP_076414.2
17
3786
NM_032156.4
NP_115532.3
18
3977
XM_006719138.3
XP_006719201.1
18
3996
XM_006719139.3
XP_006719202.1
17
3954
XM_006719140.3
XP_006719203.1
17
3951
XM_006719142.3
XP_006719205.1
17
3849
XM_006719144.3
XP_006719207.1
16
3807
XM_006719145.3
XP_006719208.1
18
4103
XM_006719146.3
XP_006719209.1
18
4135
XM_006719147.3
XP_006719210.1
18
4040
XM_017019851.1
XP_016875340.1
17
3891
XM_017019852.1
XP_016875341.1
19
3151
XM_017019853.1
XP_016875342.1
19
3221
XM_017019854.1
XP_016875343.1
16
3804
XM_017019855.1
XP_016875344.1
19
3118
XM_017019856.1
XP_016875345.1
18
3076
XM_017019857.1
XP_016875346.1
18
3046
XM_017019858.1
XP_016875347.1
18
3073
XM_017019859.1
XP_016875348.1
17
2902
XM_017019860.1
XP_016875349.1
19
4103
XM_017019861.1
XP_016875350.1
17
2926
XM_017019862.1
XP_016875351.1
19
4100
XM_017019863.1
XP_016875352.1
19
4147
XM_017019865.1
XP_016875354.1
17
3998
XM_017019866.1
XP_016875355.1
19
3198
XM_017019867.1
XP_016875356.1
19
3225
XM_017019868.1
XP_016875357.1
19
3195
XM_017019869.1
XP_016875358.1
19
3222
XM_017019870.1
XP_016875359.1
19
3135
XM_017019871.1
XP_016875360.1
19
3206
XM_017019872.1
XP_016875361.1
18
3057
XM_017019873.1
XP_016875362.1
17
2973
XM_017019874.1
XP_016875363.1
18
3068
XM_017019875.1
XP_016875364.1
17
2910
XM_017019876.1
XP_016875365.1
16
2619
XM_017019877.1
XP_016875366.1
15
2577
XM_017019878.1
XP_016875367.1
15
2574
XM_017019879.1
XP_016875368.1
17
2903
XM_017019880.1
XP_016875369.1
14
2367
XM_017019881.1
XP_016875370.1
18
3112
XM_017019882.1
XP_016875371.1
18
3052
XM_017019883.1
XP_016875372.1
15
2621
GenBank mRNA
AB058776.1
-
3423
AF525416.1
-
10
1447
AK024247.1
-
5
2902
AK026222.1
-
4
752
AL834212.1
-
16
2464
AY074490.1
-
17
3786
AY074491.1
-
17
3850
BC007520.1
-
4
657
BC066295.1
-
4
688
BC111007.1
-
4
740
BQ010043.1
-
1
562
BX537569.1
-
15
5992
BX538080.1
-
5
872
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of translation
negative regulation of cell growth
positive regulation of protein binding
positive regulation of peptidyl-serine phosphorylation
positive regulation of transcription from RNA polymerase II promoter
positive regulation of dendrite morphogenesis
positive regulation of dendritic spine morphogenesis
positive regulation of canonical Wnt signaling pathway
RNA binding
receptor binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-55874ff697-s6fgm:80/10.239.167.51:80.
git-commit: a2f5e26549390d543f5d2d1fbad8092df65fbda4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.10.0-2024.06.23-1.0