Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC28B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301011.1 | NP_001287940.1 | 3 | 600 | ||
NM_024296.4 | NP_077272.2 | 3 | 600 | |||
XM_011542113.2 | XP_011540415.1 | 3 | 1359 | |||
XM_017002306.1 | XP_016857795.1 | 3 | 600 | |||
XM_017002307.1 | XP_016857796.1 | 3 | 1354 | |||
XM_017002308.1 | XP_016857797.1 | 3 | 600 | |||
XM_017002309.1 | XP_016857798.1 | 3 | 1357 | |||
GenBank mRNA | AK225113.1 | - | 3 | 370 | ||
AK291904.1 | - | 3 | 610 | |||
BC022848.2 | - | 3 | 334 | 3946778 | ||
BM981930.1 | - | 4 | 508 |