Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THOC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142350.1 | NP_001135822.1 | 4, 5 | 615 | ||
NM_024339.3 | NP_077315.2 | 4, 5 | 615 | |||
GenBank mRNA | AK075330.1 | - | 4, 5 | 582 | ||
AK314086.1 | - | 4, 5 | 593 | |||
AL832548.1 | - | 3, 4 | 219 | |||
BC003118.1 | - | 5, 6 | 495 | 3537243 | ||
BC050674.1 | - | 4, 5 | 554 | 6067702 |