Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGBD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258311.1 | NP_001245240.1 | 3, 4 | 897 | ||
XM_017002330.1 | XP_016857819.1 | 3, 4 | 1966 | |||
GenBank mRNA | AL833243.1 | - | 2, 3 | 542 | ||
BC036865.1 | - | 2, 3 | 324 | |||
BC150638.1 | - | 3, 4 | 714 | 9057008 | ||
KF670820.1 | - | 3, 4 | 698 |