Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CCDC7 SIRNA Assays ›
Gene Symbol:
CCDC7
Gene Name:
coiled-coil domain containing 7
Gene Aliases:
BIOT2, BioT2-A, BioT2-B, BioT2-C, C10orf68
Chromosome Location:
Chr.10: 32446082 - 32882864 on Build GRCh38
UniGene ID:
Hs.585464
Species:
Human
siRNA ID
HSS128781
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CCDC7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001321115.1
NP_001308044.1
39, 40
4372
XM_005252593.2
XP_005252650.1
20, 21
2159
XM_006717505.1
XP_006717568.1
37, 38
3867
XM_006717506.1
XP_006717569.1
36, 37
3759
XM_011519663.1
XP_011517965.1
39, 40
4080
XM_011519664.1
XP_011517966.1
39, 40
4080
XM_011519666.1
XP_011517968.1
38, 39
3996
XM_011519667.1
XP_011517969.1
38, 39
3996
XM_011519668.1
XP_011517970.1
38, 39
3990
XM_011519669.1
XP_011517971.1
38, 39
3978
XM_011519670.1
XP_011517972.1
38, 39
3972
XM_011519671.1
XP_011517973.1
38, 39
3972
XM_011519672.1
XP_011517974.1
38, 39
3969
XM_011519673.1
XP_011517975.1
38, 39
3966
XM_011519674.1
XP_011517976.1
37, 38
3888
XM_011519675.1
XP_011517977.1
37, 38
3870
XM_011519676.1
XP_011517978.1
37, 38
3858
XM_011519677.1
XP_011517979.1
38, 39
3858
XM_011519678.1
XP_011517980.1
37, 38
3855
XM_011519679.1
XP_011517981.1
36, 37
3549
XM_011519680.2
XP_011517982.1
34, 35
3215
XM_011519683.2
XP_011517985.1
28, 29
2889
XM_011519685.1
XP_011517987.1
31, 32
3097
XM_011519687.1
XP_011517989.1
21, 22
2004
XM_017016638.1
XP_016872127.1
39, 40
4035
XM_017016639.1
XP_016872128.1
38, 39
3969
XM_017016640.1
XP_016872129.1
37, 38
3888
XM_017016641.1
XP_016872130.1
34, 35
3509
XM_017016642.1
XP_016872131.1
34, 35
3582
XM_017016643.1
XP_016872132.1
34, 35
3199
XM_017016645.1
XP_016872134.1
33, 34
3123
XM_017016646.1
XP_016872135.1
30, 31
3010
XM_017016648.1
XP_016872137.1
26, 27
2731
XM_017016649.1
XP_016872138.1
26, 27
2671
XM_017016652.1
XP_016872141.1
23, 24
2134
XM_017016653.1
XP_016872142.1
21, 22
2044
GenBank mRNA
AK023093.1
-
18, 19
1973
AK097668.1
-
12, 13
1113
AK301824.1
-
17, 18
1555
BC125090.1
-
19, 20
1995
40118176
BC125091.1
-
17, 18
1582
40118177
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-r642z:80/10.239.167.51:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0