Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EHMT1 SIRNA Assays ›
Gene Symbol:
EHMT1
LOC651337
Gene Name:
euchromatic histone lysine methyltransferase 1
uncharacterized LOC651337
Gene Aliases:
EUHMTASE1, Eu-HMTase1, FP13812, GLP, GLP1, KMT1D, bA188C12.1
-
Chromosome Location:
Chr.9: 137618988 - 137836127 on Build GRCh38
Chromosome Location:
-
UniGene ID:
Hs.495511
Hs.603195
Species:
Human
siRNA ID
HSS128909
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EHMT1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145527.1
NP_001138999.1
12
1902
NM_024757.4
NP_079033.4
12
1902
XM_005266105.4
XP_005266162.1
12
1945
XM_005266110.1
XP_005266167.1
11
1817
XM_006717288.2
XP_006717351.1
12
1863
XM_011519021.2
XP_011517323.1
12
2900
XM_011519022.2
XP_011517324.1
12
2355
XM_011519023.2
XP_011517325.1
12
2879
XM_011519024.2
XP_011517326.1
11
2823
XM_011519025.1
XP_011517327.1
12
2065
XM_011519026.2
XP_011517328.1
11
2758
XM_011519027.2
XP_011517329.1
12
2901
XM_011519028.2
XP_011517330.1
12
2901
XM_011519029.2
XP_011517331.1
4
490
XM_011519033.2
XP_011517335.1
11
2737
XM_017015134.1
XP_016870623.1
12
2333
XM_017015135.1
XP_016870624.1
12
1885
XM_017015136.1
XP_016870625.1
11
1807
XM_017015137.1
XP_016870626.1
12
2044
XM_017015138.1
XP_016870627.1
11
1796
GenBank mRNA
AB028932.1
-
7
722
AB058779.2
-
7
722
AK022941.1
-
11
1817
AY083210.1
-
10
1772
BC011608.2
-
2
119
BC047504.1
-
12
1902
5298029
Gene Symbol: LOC651337
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
GenBank mRNA
AK124119.1
-
2
1755
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
DNA methylation
embryo development
chromatin modification
histone methylation
peptidyl-lysine monomethylation
peptidyl-lysine dimethylation
negative regulation of transcription, DNA-templated
histone H3-K9 methylation
response to fungicide
histone H3-K27 methylation
regulation of signal transduction by p53 class mediator
p53 binding
protein binding
methyltransferase activity
zinc ion binding
protein-lysine N-methyltransferase activity
histone-lysine N-methyltransferase activity
histone methyltransferase activity (H3-K9 specific)
histone methyltransferase activity (H3-K27 specific)
C2H2 zinc finger domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-jhld4:80/10.239.165.190:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0