Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167941.1 | NP_001161413.1 | 4 | 594 | ||
NM_024908.3 | NP_079184.2 | 4 | 704 | |||
XM_005254680.3 | XP_005254737.1 | 4 | 667 | |||
GenBank mRNA | AK023035.1 | - | 4 | 609 | ||
AK290933.1 | - | 4 | 626 | |||
BC025247.1 | - | 4 | 635 | 4560352 | ||
BC051855.1 | - | 4 | 607 | 6141090 | ||
BX374706.2 | - | 4 | 520 | |||
CN259143.1 | - | 4 | 645 | |||
EF011624.1 | - | 4 | 607 |