Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195059.1 | NP_001181988.1 | 2, 3 | 265 | ||
NM_025201.4 | NP_079477.2 | 3, 4 | 415 | |||
GenBank mRNA | AF318373.1 | - | 13 | |||
AF494534.1 | - | 3, 4 | 364 | |||
AK096203.1 | - | 3, 4 | 383 | |||
BC008744.2 | - | 2, 3 | 226 | 3346334 |