Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030773.3 | NP_110400.1 | 1, 2 | 332 | ||
GenBank mRNA | AJ292757.1 | - | 1, 2 | 332 | ||
BC033679.1 | - | 1, 2 | 165 | 5180793 | ||
BI821931.1 | - | 1, 2 | 165 |