Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015059.2 | NP_055874.2 | 18 | 2414 | ||
XM_005254708.4 | XP_005254765.1 | 21 | 2782 | |||
XM_005254710.4 | XP_005254767.1 | 22 | 2920 | |||
XM_005254711.4 | XP_005254768.1 | 21 | 3142 | |||
XM_005254712.4 | XP_005254769.1 | 21 | 2799 | |||
XM_005254713.4 | XP_005254770.1 | 21 | 3696 | |||
XM_005254714.3 | XP_005254771.1 | 20 | 2685 | |||
XM_005254715.2 | XP_005254772.1 | 20 | 3391 | |||
XM_006720717.3 | XP_006720780.1 | 21 | 3516 | |||
XM_017022665.1 | XP_016878154.1 | 21 | 3867 | |||
XM_017022666.1 | XP_016878155.1 | 21 | 2726 | |||
XM_017022667.1 | XP_016878156.1 | 19 | 2588 | |||
XM_017022668.1 | XP_016878157.1 | 21 | 3515 | |||
XM_017022669.1 | XP_016878158.1 | 21 | 3516 | |||
GenBank mRNA | AB002318.2 | - | 4 | 588 | ||
AF402000.1 | - | 18 | 2414 |