Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303096.1 | NP_001290025.1 | 12 | 1188 | ||
NM_032168.2 | NP_115544.1 | 11 | 1154 | |||
GenBank mRNA | AK022581.1 | - | 11 | 1101 | ||
AK022607.1 | - | 11 | 1113 | |||
AK023276.1 | - | 8 | 793 | |||
AK091546.1 | - | 12 | 1115 | |||
AK290445.1 | - | 11 | 1105 | |||
AL122097.1 | - | 7 | 721 | |||
BC040567.1 | - | 11 | 1154 | 5267120 | ||
CR749440.1 | - | 11 | 2763 |