Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MON1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032355.3 | NP_115731.2 | 3 | 882 | ||
XM_006713345.3 | XP_006713408.1 | 3 | 648 | |||
XM_011534160.1 | XP_011532462.1 | 3 | 511 | |||
GenBank mRNA | AK074404.1 | - | 3 | 692 | ||
AK315630.1 | - | 3 | 736 | |||
BC006299.2 | - | 3 | 694 | 4079816 | ||
BC009459.1 | - | 3 | 651 | 3536968 |