Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130783.4 | NP_570139.3 | 3 | 267 | ||
XM_005253217.3 | XP_005253274.1 | 5 | 451 | |||
XM_006718372.3 | XP_006718435.1 | 5 | 538 | |||
XM_006718373.3 | XP_006718436.1 | 4 | 524 | |||
XM_011520459.2 | XP_011518761.1 | 4 | 414 | |||
XM_017018530.1 | XP_016874019.1 | 5 | 484 | |||
XM_017018531.1 | XP_016874020.1 | 4 | 406 | |||
GenBank mRNA | AK027715.1 | - | 2 | 76 | ||
AY358087.1 | - | 2 | 190 | |||
BC019342.2 | - | 1 | 39 | 3357734 | ||
DA734054.1 | - | 4 | 317 |