Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2Q2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145335.1 | NP_001138807.1 | 7 | 972 | ||
NM_001284382.1 | NP_001271311.1 | 6 | 986 | |||
NM_173469.3 | NP_775740.1 | 7 | 1091 | |||
XM_005254787.1 | XP_005254844.1 | 7 | 863 | |||
XM_017022726.1 | XP_016878215.1 | 7 | 967 | |||
XM_017022727.1 | XP_016878216.1 | 6 | 786 | |||
GenBank mRNA | AK000617.1 | - | 6 | 965 | ||
AK294199.1 | - | 7 | 632 | |||
AK296234.1 | - | 7 | 972 | |||
AL832429.1 | - | 7 | 916 | |||
BC006827.1 | - | 5 | 366 | |||
BC017708.1 | - | 7 | 918 | 3907760 |