Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CADPS2 SIRNA Assays ›
Gene Symbol:
CADPS2
Gene Name:
calcium dependent secretion activator 2
Gene Aliases:
CAPS2
Chromosome Location:
Chr.7: 122318424 - 122886759 on Build GRCh38
UniGene ID:
Hs.649459
Species:
Human
siRNA ID
HSS132566
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CADPS2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001009571.3
NP_001009571.2
7
1694
NM_001167940.1
NP_001161412.1
7
1694
NM_017954.10
NP_060424.9
7
1694
XM_005250695.4
XP_005250752.1
7
1423
XM_005250696.4
XP_005250753.1
7
1423
XM_005250697.4
XP_005250754.1
7
1423
XM_005250698.4
XP_005250755.1
7
1395
XM_005250699.4
XP_005250756.1
7
1423
XM_005250700.4
XP_005250757.1
7
1423
XM_005250701.4
XP_005250758.1
7
1423
XM_005250702.4
XP_005250759.1
7
1423
XM_005250703.4
XP_005250760.1
7
1423
XM_005250704.4
XP_005250761.1
7
1423
XM_005250705.4
XP_005250762.1
7
1423
XM_005250706.4
XP_005250763.1
7
1423
XM_005250707.4
XP_005250764.1
7
1423
XM_005250708.3
XP_005250765.1
7
1423
XM_011516695.2
XP_011514997.1
7
1395
XM_011516696.2
XP_011514998.1
7
1423
XM_011516697.2
XP_011514999.1
7
1423
XM_017012794.1
XP_016868283.1
7
1423
XM_017012795.1
XP_016868284.1
7
1423
XM_017012796.1
XP_016868285.1
8
1038
XM_017012797.1
XP_016868286.1
8
1104
XM_017012798.1
XP_016868287.1
7
1574
XM_017012799.1
XP_016868288.1
8
1475
XM_017012800.1
XP_016868289.1
4
431
GenBank mRNA
AB046811.1
-
5
563
AF401638.1
-
7
1363
AK025672.1
-
3
217
AK126518.1
-
8
1296
AL833058.1
-
4
489
AY264289.1
-
7
1298
BC136601.1
-
7
1393
9020591
BC144278.1
-
7
1393
9052798
BC144279.1
-
7
1393
9052799
KJ534798.1
-
7
1272
KJ534800.1
-
7
1272
KJ534801.1
-
7
1272
KJ534802.1
-
7
1272
KJ534803.1
-
7
1272
KJ534804.1
-
7
1272
KJ534805.1
-
7
1272
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane trafficking regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
cellular response to starvation
protein transport
synaptic vesicle priming
positive regulation of exocytosis
dense core granule exocytosis
lipid binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-zp8kv:80/10.239.163.191:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0